H-His-Leu-OH
Supplier: BACHEM AMERICAS INC
4000307.0001 4000307.0005
G-2310.0001BAEA 464.74 CAD
G-2310.0001BA G-2310.0005BA
H-His-Leu-OH
Proteins and Peptides
The sequences given for angiotensin I (H-1680) and II (H-1705) correspond to the sequences of human, horse, sheep, pig, mouse, and rat angiotensins I and II.
His-Leu, C-terminal dipeptide of angiotensin I is split off by action of angiotensin-converting enzyme (ACE) yielding angiotensin II. See also M-1305 and M-1485. The complex generated from Cu(II), His-Leu and ethylene diamine (or His) hydrolytically cleaves the phosphodiester backbone of DNA.