Glycylsarcosine

Supplier: BACHEM AMERICAS INC

Synonyms: H-Gly-Sar-OH

4001240.0005 4001240.0025
G-2195.0005BAEA 317.63 CAD
G-2195.0005BA G-2195.0025BA
Glycylsarcosine
Glycylsarcosine

Due to its resistance to hydrolysis, glycylsarcosine served as a model dipeptide used to study peptide transport in a variety of cell lines.

Formula: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038193
CAS Number: 29816-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR