H-Gly-Phe-OH
Supplier: BACHEM AMERICAS INC
4000165.0005 4000165.0025
G-2175.0005BAEA 211.77 CAD
G-2175.0005BA G-2175.0025BA
H-Gly-Phe-OH
Proteins and Peptides
A bitter-tasting dipeptide, which activates the human bitter receptors hTAS2Rs.