H-Gly-His-OH
Supplier: BACHEM AMERICAS INC
4000976.0001 4000976.0005
G-2120.0001BAEA 516.34 CAD
G-2120.0001BA G-2120.0005BA
H-Gly-His-OH
Proteins and Peptides
Short histidine-containing peptides can form stable complexes with copper and other transition metals. See also His-Leu (G-2310) and the product family 'Liver Cell Growth Factor (GHK) and Analogs'.
The copper-complexing dipeptide glycyl-L-histidine also forms complexes with Au (III) and Pd (II).