H-Gly-Gly-OH
Supplier: BACHEM AMERICAS INC
4030664.0025
4030664.0100
G-2090.0025BAEA
180.91
CAD
G-2090.0025BA
G-2090.0100BA
H-Gly-Gly-OH
Proteins and Peptides
Standard acceptor in γ-glutamyltransferase assays, e.g. with H-Glu(pNA)-OH (L-1265) or H-Glu(AMC)-OH (I-1185).