Human beta-Endorphin (30-31)
Supplier: BACHEM AMERICAS INC
4002438.0005 4002438.0025
G-2080.0005BAEA 284.04 CAD
G-2080.0005BA G-2080.0025BA
Human beta-Endorphin (30-31)
Proteins and Peptides
For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Dynorphin, Analogs and Sequences, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides’.
The C-terminal dipeptide Gly-Glu was shown to be an effective neurotrophic factor for the maintenance of acetylcholinesterase at both superior cervical ganglia in the denervated cat. Moreover, β-endorphin can bind to serum glycoproteins at sites for which glycyl-L-glutamic acid competes. A potential impurity of glycyl-L-glutamine used as source for L-glutamine.