H-Glu(Val-OH)-OH
Supplier: BACHEM AMERICAS INC
4003707.0250 4003707.1000
G-2015.0250BAEA 877.75 CAD
G-2015.0250BA G-2015.1000BA
H-Glu(Val-OH)-OH
Proteins and Peptides
See also G-4745, H-3996, the branched substrates H-2416, and L-2070, and the product families 'Glutathione-Related Peptides' and 'Ophthalmic Acid'.
γ-Glu-Val is a main contributor to the "kokumi" taste of edible beans (Phaseolus vulgaris L.)