H-Glu(Tyr-OH)-OH
Supplier: BACHEM AMERICAS INC
4002026.0250 4002026.1000
G-2005.0250BAEA 476.37 CAD
G-2005.0250BA G-2005.1000BA
H-Glu(Tyr-OH)-OH
Proteins and Peptides
See also G-4745, H-3996, the branched substrates H-2416, and L-2070, and the product families 'Glutathione-Related Peptides' and 'Ophthalmic Acid'.
The γ-glutamyl dipeptides γ-Glu-Tyr, γ-Glu-Leu (G-1950) and γ-Glu-Phe (G-1975) have been detected in Comté cheese. In combination, they probably contribute to the complex typical flavor of this and other types of hard cheese. The soluble γ-glutamyltyrosine can serve as tyrosine source.