H-Glu-Trp-OH
Supplier: BACHEM AMERICAS INC
4000564.0250 4000564.1000
G-1990.0250BAEA 439.41 CAD
G-1990.0250BA G-1990.1000BA
H-Glu-Trp-OH
Proteins and Peptides
See also the product families 'Adjuvant Peptides' and 'Thymosins and Fragments'.
A naturally occurring thymic immunomodulator. EW in vivo regulates NK cell function independent of IFNγ but partly dependent on IL-12. Substrate for aminopeptidase W, Km 0.49 mM and kcat 5557 min⁻¹ .