H-Glu(Phe-OH)-OH
Supplier: BACHEM AMERICAS INC
4002375.0250 4002375.1000
G-1975.0250BAEA 413.58 CAD
G-1975.0250BA G-1975.1000BA
H-Glu(Phe-OH)-OH
Proteins and Peptides
See also G-4745, H-3996, the branched substrates H-2416, and L-2070, and the product families 'Glutathione-Related Peptides' and 'Ophthalmic Acid'.
This branched dipeptide was detected in the urine of patients with phenylketonuria and later on in the urine of healthy individuals. γ-Glu-Phe and other branched dipeptides have been isolated from hard cheese, where they contribute to the typical flavor of this dairy product.