H-Glu-Glu-OH
Supplier: BACHEM AMERICAS INC
4000466.0250
4000466.1000
G-1915.0250BAEA
232.35
CAD
G-1915.0250BA
G-1915.1000BA
H-Glu-Glu-OH
Proteins and Peptides
Dipeptides as aspartylaspartate (G-1565) and glutamylglutamate can be utilized as growth substrates for the oral bacteria Porphyromonas gingivalis, Prevotella intermedia, Prevotella nigrescens and Fusobacterium nucleatum.