Cyclo(-Asp-Phe)
Supplier: BACHEM AMERICAS INC
4016611.0250
4016611.1000
G-1695.0250BAEA
1161.77
CAD
G-1695.0250BA
G-1695.1000BA
Cyclo(-Asp-Phe)
Proteins and Peptides
The diketopiperazine cyclo(Asp-Phe) is the major degradation product of aspartame in food, e.g. during heating. Hence, the pure material is an important reference substance for the analysis of foodstuffs containing this sweetener.