H-Asp-Asp-OH, Bachem
Supplier: BACHEM AMERICAS INC
4010210.0250
4010210.1000
G-1565.0250BAEA
317.42
CAD
G-1565.0250BA
G-1565.1000BA
H-Asp-Asp-OH, Bachem
Proteins and Peptides
Dipeptides as aspartylaspartate and glutamylglutamate (G-1915) can be utilized as growth substrates for the oral bacteria Porphyromonas gingivalis, Prevotella intermedia, Prevotella nigrescens and Fusobacterium nucleatum.