H-Arg-Arg-OH acetate, Bachem
Supplier: BACHEM AMERICAS INC
4000091.0250 4000091.1000
G-1465.0250BAEA 722.85 CAD
G-1465.0250BA G-1465.1000BA
H-Arg-Arg-OH acetate, Bachem
Proteins and Peptides
Arginine-containing dipeptides as RR and RF (G-1515) are used by nitric oxide synthase as substrates in place of Arg.