L-Alanyl-L-tyrosine

Supplier: BACHEM AMERICAS INC

Synonyms: H-Ala-Tyr-OH, (S)-2-((S)-2-Aminopropanamido)-3-(4-hydroxyphenyl)propanoic acid, L-Alanyl-L-tyrosine

4005177.0001 4005177.0005
G-1405.0001BAEA 529.25 CAD
G-1405.0001BA G-1405.0005BA
L-Alanyl-L-tyrosine
L-Alanyl-L-tyrosine

Sequence: H-Ala-Tyr-OH


These short peptides act as angiotensin I-converting enzyme (ACE) inhibitors. See also Phe-Tyr (G-2955) and the subfamilies 'Bradykinin-Potentiating Peptide (BPP)' and 'Angiotensin I-Converting Enzyme (ACE, ACE2) Inhibitors'.


Inhibitor of angiotensin-1 converting enzyme (ACE), IC₅₀ 14.2 μM. Ala-Tyr has been used as a tyrosine source in intravenous nutrition of the rat. The dipeptide AY is an efficient Tyr source for the parenteral nutrition of patients with hepatic failure.

Formula: C₁₂H₁₆N₂O₄
MW: 252.27 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038164
CAS Number: 3061-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR