H-Ala-D-Glu-NH₂, Bachem

Supplier: BACHEM AMERICAS INC

4007604.0250 4007604.1000
G-1255.0250BAEA 570.65 CAD
G-1255.0250BA G-1255.1000BA
H-Ala-D-Glu-NH₂, Bachem
Proteins and Peptides

Partially protected Ala-D-IsoGln and Ala-IsoGln dipeptides.

Order Now

SPECIFICATIONS

Protein/peptide name H-Ala-D-Glu-NH₂
Environmentally Preferable
Protein synonyms H-Ala-D-Isogln-OH
Species
Source
CAS No. 45159-25-9
MDL No. MFCD00237727
Sequence H-Ala-D-Glu-NH₂
Molecular weight 217.23
Molecular formula C₈H₁₅N₃O₄
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR