L-Carnosine
Supplier: BACHEM AMERICAS INC
4030364.0005
4030364.0025
G-1250.0005BAEA
500.88
CAD
G-1250.0005BA
G-1250.0025BA
L-Carnosine
Proteins and Peptides
Carnosine is a major component of the non-protein fraction of vertebrate skeletal muscle. It acts as an aldehyde scavenger, a likely lipofuscin (age pigment) precursor and possible modulator of diabetic complications, atherosclerosis and Alzheimer's disease. Janssen et al. showed that carnosine acts as a protective factor in diabetic nephropathy.