H-Ala-Gln-OH, Bachem
Supplier: BACHEM AMERICAS INC
4003766.0001
4003766.0005
G-1210.0001BAEA
697.9
CAD
G-1210.0001BA
G-1210.0005BA
H-Ala-Gln-OH, Bachem
Proteins and Peptides
The dipeptides H-Ala-Gln-OH and H-Gly-Gln-OH (N-1070) are stable substitutes for Gln in cell culture media, which tolerate autoclaving and liberate a lower amount of undesired ammonia than Gln. Ala-Gln and Gly-Gln have also found use as Gln sources in parenteral nutrition.