H-Ala-Ala-OH, Bachem
Supplier: BACHEM AMERICAS INC
4004429.0005 4004429.0025
G-1120.0005BAEA 542.79 CAD
G-1120.0005BA G-1120.0025BA
H-Ala-Ala-OH, Bachem
Proteins and Peptides
Substrate for a fluorometric assay of urinary dipeptidase activity. Ala-Ala (dialanine) was found to inhibit the rooting of germinating seeds. Ala-Ala acts as an efficient enatioselective catalyst in aldol reactions.