Ac-(6-O-stearoyl)-muramyl-Ala-D-Glu-NH₂
Supplier: BACHEM AMERICAS INC
4015905.0001 4015905.0005
G-1065.0001BAEA 479.83 CAD
G-1065.0001BA G-1065.0005BA
Ac-(6-O-stearoyl)-muramyl-Ala-D-Glu-NH₂
Proteins and Peptides
The lipidated L18 MDP showed a higher activity than unmodified MDP. L18 MDP acts as a NOD2 receptor agonist.