Ac-Asp-Glu-OH

Supplier: BACHEM AMERICAS INC

4006232.0100 4006232.0250
G-1015.0100BAEA 387.21 CAD
G-1015.0100BA G-1015.0250BA
Ac-Asp-Glu-OH
Proteins and Peptides

An endogenous dipeptide with high affinity for a brain glutamate receptor. NAAG is the most abundant peptide neurotransmitter in the mammalian central nervous system.

Order Now

SPECIFICATIONS

Protein/peptide name Ac-Asp-Glu-OH
Protein synonyms Spaglumic acid|NAAG
Environmentally Preferable
Species
Source
CAS No. 3106-85-2
MDL No. MFCD00076652
Sequence Ac-Asp-Glu-OH
Molecular weight 304.26
Molecular formula C₁₁H₁₆N₂O₈
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR