Sodium pyrophosphate, 200 mM buffer solution., Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62052-AE
J62052-AK
CAAAJ62052-AEEA
150.07
CAD
CAAAJ62052-AE
CAAAJ62052-AK
Sodium pyrophosphate, 200 mM buffer solution., Thermo Scientific
Buffers
Cloning Buffers
Liquid
CAS No.: 7758-16-9
Physical Form: Liquid
MDL No.: MFCD00014246
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...