Sodium Acetate, 3M aqua solution, pH 5.2, RNAse free, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61928-AK J61928-AE
CAAAJ61928-AKEA 225.04 CAD
CAAAJ61928-AK CAAAJ61928-EQE AAJ61928-AE
Sodium Acetate, 3M aqua solution, pH 5.2, RNAse free, Thermo Scientific
Buffers Cloning Buffers

Sodium acetate buffer solution pH, 5.2 is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as Chip-on-Chip assay and in vitro enzyme inhibition assay.


  • Sodium acetate buffer solution pH, 5.2 is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as Chip-on-Chip assay and in vitro enzyme inhibition assay.
  • Soluble in water (246 g/L) at 20°C, and ethanol.
  • Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, halogens. Moisture sensitive. Stable under recommended storage conditions. Keep away from strong oxidizing agent. .


CAS No.: 127-09-3
Molecular Formula: C₂H₃NaO₂.
Formula Weight: 82.03 g/mol
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00012459


Caution: RUO – Research Use Only

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR