Potassium iodide 15% w/v aqueous solution Stabilised

Supplier: Ricca Chemical
Danger

Synonyms: Dipotassium diiodide, Kali iodide

R6305000-500B R6305000-1B
RC630516EA 202.24 CAD
RC630516 RC630532
Potassium iodide 15% w/v aqueous solution Stabilised
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686
Density: 1.1086 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
Merck Index: 13,07727

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 15.0 ± 0.1 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR