H-Arg-Val-Leu-psi(CH₂NH)Phe-Glu-Ala-Nle-NH₂
Supplier: BACHEM AMERICAS INC
4030746.0001
N-1270.0001BAEA 1395.84 CAD
N-1270.0001BA
H-Arg-Val-Leu-psi(CH₂NH)Phe-Glu-Ala-Nle-NH₂
Proteins and Peptides
See also Ac-pepstatin (N-1250).
The peptide RVL-psi[CH2NH]FEA-Nle-amide containing a reduced peptide bond was found to be a competitive inhibitor of the HIV-1 protease (Ki = 50 nM).