Methyl L-tryptophanate hydrochloride

Supplier: BACHEM AMERICAS INC

4000133.0025 4000133.0100
N-1165.0025BAEA 490.51 CAD
N-1165.0025BA N-1165.0100BA
Methyl L-tryptophanate hydrochloride
Methyl L-tryptophanate hydrochloride

Sequence: H-Trp-OMe · HCl


A competitive inhibitor of dexamethasone binding to the glucocorticoid receptor in HTC cells.

Formula: C₁₂H₁₄N₂O₂·HCl
Storage Temperature: Freezer
MDL Number: MFCD00066134
CAS Number: 7524-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR