Masterflex® Miniflex® OEM Panel Mount Pumps Systems
Supplier: Avantor Fluid Handling
77222-00 77222-02 77222-10 77222-12 77222-14
MFLX77222-00EA 2305.22 CAD
MFLX77222-00 MFLX77222-02 MFLX77222-10 MFLX77222-12 MFLX77222-14
Masterflex® Miniflex® OEM Panel Mount Pumps Systems
Pumps Peristaltic Pumps
Designed for easy integration into OEM designs wherever precision fluid handling is needed.
  • Precise flowrates up from 0.12 to 390 ml/min (3 to 300 rpm)
  • Rugged sturdy design with durable pump head materials
  • Pump head accepts continuous lengths of tubing for clean flow paths
  • Automatic tubing retention makes tube loading quick and easy
  • IP 56 Ingress protection; removable head for spray and wipe down cleanability
  • High efficiency and dependable brushless DC motor
  • I/O control features include analog speed control, direction, tach output, open head sensor alarm,
More Product Information