DRIVE PANEL MOUNT MINIFLEX 4-20MA 24VDC
Supplier: Avantor Fluid Handling
VWR Item#MFLX77222-00
Your Organization's Price:
Retrieving
Designed for easy integration into OEM designs wherever precision fluid handling is needed.
  • <p>Precise flowrates up from 0.12 to 390 ml/min (3 to 300 rpm)<br />
  • Rugged sturdy design with durable pump head materials<br />
  • Pump head accepts continuous lengths of tubing for clean flow paths<br />
  • Automatic tubing retention makes tube loading quick and easy<br />
  • IP 56 Ingress protection; removable head for spray and wipe down cleanability<br />
  • High efficiency and dependable brushless DC motor<br />
  • I/O control features include analog speed control, direction, tach output, open head sensor alarm, and general alarm<br />
  • Reduced pulsation options available with 4 or 6 roller pump heads<br />
  • Multichannel capability with dual channel pump head option<br />
  • More than 20 tubing formulations to support chemical and compliance requirements<br />
  • Customizable options available</p>
Applications: Biopharma production, testing and research; Laboratory instrumentation; Air and liquid sampling devices; Food and beverage dispensing; Ink and printing; Industrial fluid transfer; Wastewater management.
Miniflex® panel mount systems include everything needed to incorporate Masterflex® peristaltic pumping into OEM product designs. These models feature an IP65 ingress protection to ensure proper seal during cleaning and for use in critical environments. The BLDC motors provide a combination of efficiency and long term durability. Miniflex® pump head allows for fast tubing changes with up to four tubing sizes.
<b>Certifications: </b>UL 61010-1, CSA, CE (EN 61010-1 Low Voltage, EN 61326-1 EMC Directive, EN ICE 63000 ROSH Directive, EN 809 Machinery Directive), ISO 9001.
<b>Ordering information: </b>For use with Masterflex® Pump tubing, sold separately. Two channel pump head options are available and pump system is rated for use with all Masterflex® tubing formulations. Choose four roller pump head design for reduced pulsation. Customized options are available.
<b>Delivery information: </b>Includes pump drive, power and IO cable, pump heads sold separately.