Leu-Leu

Supplier: BACHEM AMERICAS INC

4001608.0005 4001608.0025
M-1535.0005BAEA 922.99 CAD
M-1535.0005BA M-1535.0025BA
Leu-Leu
Leu-Leu

Sequence: H-Leu-Leu-OH


Dileucine, substrate for dipeptidase activity measurement in serum and tissues as well as urine. LL forms nanotubes.

Formula: C₁₂H₂₄N₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00065185
CAS Number: 3303-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR