Glycyl-L-leucine

Supplier: BACHEM AMERICAS INC

4000534.0025 4000534.0100
M-1460.0025BAEA 516.34 CAD
M-1460.0025BA M-1460.0100BA
Glycyl-L-leucine
Glycyl-L-leucine

Sequence: H-Gly-Leu-OH


A bitter-tasting dipeptide, which activates the human bitter receptors hTAS2Rs. GL is a good substrate for dipeptide hydrolase activities in rat intestinal mucosa and human kidney.

Formula: C₈H₁₆N₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00008127
CAS Number: 869-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR