Z-Gly-Pro-Gly-Gly-Pro-Ala-OH

Supplier: BACHEM AMERICAS INC

4002149.0100 4002149.0500
M-1260.0100BAEA 185.3 CAD
M-1260.0100BA M-1260.0500BA
Z-Gly-Pro-Gly-Gly-Pro-Ala-OH
Z-Gly-Pro-Gly-Gly-Pro-Ala-OH

Sequence: Z-Gly-Pro-Gly-Gly-Pro-Ala-OH


Substrate for collagenase. Enzymatic cleavage of Z-GPGGPA yields Gly-Pro-Ala (H-3615), which is determined colorimetrically by reaction with ninhydrin.

Formula: C₂₇H₃₆N₆O₉
Storage Temperature: Freezer
MDL Number: MFCD00077021
CAS Number: 13075-38-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR