For-Met-Leu-pNA
Supplier: BACHEM AMERICAS INC
4027984.0250
L-2030.0250BAEA 1861.45 CAD
L-2030.0250BA
For-Met-Leu-pNA
Proteins and Peptides
fML-pNA, chromogenic substrate for a continuous spectrophotometric assay for peptide deformylase, see also L-2055.