Suc-Ala-Ala-Val-pNA
Supplier: BACHEM AMERICAS INC
4003470.0050
L-1405.0050BAEA 258.14 CAD
L-1405.0050BA
Suc-Ala-Ala-Val-pNA
Proteins and Peptides
See also C-1040, C-2345, I-1000, and L-1435.
Suc-AAV-pNA (SAAVNA), a very specific substrate for human and rat leukocyte elastases.