(S)-N-(2-Aminoacetyl)-1-(4-nitrophenyl)pyrrolidine-2-carboxamide-4-methylbenzenesulfonate

Supplier: BACHEM AMERICAS INC

4003654.0100 4003654.0250
L-1295.0100BAEA 335.58 CAD
L-1295.0100BA L-1295.0250BA
(S)-N-(2-Aminoacetyl)-1-(4-nitrophenyl)pyrrolidine-2-carboxamide-4-methylbenzenesulfonate
(S)-N-(2-Aminoacetyl)-1-(4-nitrophenyl)pyrrolidine-2-carboxamide-4-methylbenzenesulfonate

Sequence: H-Gly-Pro-pNA · p-tosylate


See also H-Lys-Pro-AMC (I-1745) and H-Lys-Pro-MNA (J-1255) .


Very good substrate for the assay of dipeptidyl-peptidase IV (DPP IV, X-prolyldipeptidyl aminopeptidase), especially in normal and pathological human sera.

Formula: C₂₀H₂₄N₄O₇S
Storage Temperature: Freezer
MDL Number: MFCD00039007
CAS Number: 65096-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR