Z-Arg-Arg-pNA·2 HCl

Supplier: BACHEM AMERICAS INC

4008026.0050 4008026.0250
L-1225.0050BAEA 455.91 CAD
L-1225.0050BA L-1225.0250BA
Z-Arg-Arg-pNA·2 HCl
Proteins and Peptides

Z-RR-pNA, chromogenic substrate for cathepsin B and stem bromelain.

Order Now

SPECIFICATIONS

Protein/peptide name Z-Arg-Arg-pNA · 2 HCl
Environmentally Preferable
Species
Source
CAS No. 201807-90-1
MDL No. MFCD00155553
Sequence Z-Arg-Arg-pNA · 2 HCl
Molecular weight 657.56
Molecular formula C₂₆H₃₆N₁₀O₆ · 2HCl
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR