H-Gly-Trp-β-NA·HCl

Supplier: BACHEM AMERICAS INC

4002635.0250 4002635.1000
K-1345.0250BAEA 903.62 CAD
K-1345.0250BA K-1345.1000BA
H-Gly-Trp-β-NA·HCl
H-Gly-Trp-beta-NA·HCl

Sequence: H-Gly-Trp-βNA · HCl

Formula: C₂₃H₂₃ClN₄O₂
Storage Temperature: Freezer
MDL Number: MFCD02259641
CAS Number: 370564-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR