H-Ala-Ala-βNA

Supplier: BACHEM AMERICAS INC

4002328.0250 4002328.1000
K-1040.0250BAEA 309.81 CAD
K-1040.0250BA K-1040.1000BA
H-Ala-Ala-βNA
H-Ala-Ala-betaNA

Sequence: H-Ala-Ala-βNA


A good substrate for dipeptidyl aminopeptidase I (cathepsin C).

Formula: C₁₆H₁₉N₃O₂
Storage Temperature: Freezer
MDL Number: MFCD00151974
CAS Number: 20724-07-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR