Agarose, Low Melting Point, IBI Scientific
Supplier: IBI Scientific
IB70051 IB70056 IB70057 IB70058 IB70059 IB70050
89125-532EA 233.78 CAD
89125-532 89125-534 89125-536 89133-104 89133-106 IB70050
Agarose, Low Melting Point, IBI Scientific
Electrophoresis Gels Agarose and Acrylamide
Low Melting Point Agarose is commonly used as a gel matrix when fragment recovery is desired after electrophoresis
Some features of IBI Low Melt Agarose when compared to standard agarose are higher clarity, greater sieving capacity, and lower gel strength yet still easy to handle.
IBI Low Melt Agarose can be used for DNA electrophoresis, RNA electrophoresis, cloning of tissue culture cells, and viral plague assays. Low Melt Agarose gels will separate and finely resolve molecules that are > 1000bp.
IBI Low Melt Agarose is derived by organic synthesis, which generates methoxylateMore Product Information