Z-Ala-Ala-ASN-AMC

Supplier: BACHEM AMERICAS INC

4033201.0050 4033201.0250
I-1865.0050BAEA 472.42 CAD
I-1865.0050BA I-1865.0250BA
Z-Ala-Ala-ASN-AMC
Z-Ala-Ala-ASN-AMC

Sequence: Z-Ala-Ala-Asn-AMC


Z-AAN-AMC, standard substrate for legumain (EC 3.4.22.34), was originally used for the plant enzyme and later to discover the mammalian form of the enzyme. Legumain may be a physiologic local regulator of human osteoclast (OCL) activity that can negatively modulate OCL.

Formula: C₂₈H₃₁N₅O₈
Storage Temperature: Freezer
MDL Number: MFCD02684274
CAS Number: 149697-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR