Human;Pig;Rat (Met(O)²⁷)-Glucagon (1-29) Trifluoroacetate

Supplier: BACHEM AMERICAS INC

4043506.0500 4043506.1000
H-6148.0500BAEA 645.43 CAD
H-6148.0500BA H-6148.1000BA
Human;Pig;Rat (Met(O)²⁷)-Glucagon (1-29) Trifluoroacetate
Proteins and Peptides

For the long-acting GLP-1 analog liraglutide see H-6724.


This glucagon derivative shows the same maximal glucose mobilizing activity in rat hepatocytes as native glucagon, but it is less potent, suggesting a crucial role of methionine in the binding of glucagon to its hepatic receptor.

Order Now

SPECIFICATIONS

Protein/peptide name (Met(O)²⁷)-Glucagon (1-29) Trifluoroacetate
Environmentally Preferable
Species Pig,Human,Rat
Source
CAS No. 75217-63-9
MDL No. MFCD18782362
Sequence H-His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met(O)-Asn-Thr-OH
Molecular weight 3498.80
Molecular formula C₁₅₃H₂₂₅N₄₃O₅₀S
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR