H-Ser-Ser-Ser-OH
Supplier: BACHEM AMERICAS INC
4002153.0250 4002153.1000
H-4980.0250BAEA 722.85 CAD
H-4980.0250BA H-4980.1000BA
H-Ser-Ser-Ser-OH
Proteins and Peptides
Triserine, H-4980, was shown to increase the release of vasodilatory substances as PGI₂ and NO.