(Phe²,Orn⁸)-Oxytocin
Supplier: BACHEM AMERICAS INC
4030889.0005 4030889.0025
H-3178.0005BAEA 697.9 CAD
H-3178.0005BA H-3178.0025BA
(Phe²,Orn⁸)-Oxytocin
Proteins and Peptides
For atosiban see H-6722.
Potent vasopressor (V₁) agonist with very little antidiuretic (V₂) activity.