Bovine;Human;Pig Thr-Ala-Pro-Arg-Atrial Natriuretic Factor (1-28) Trifluoroacetate

Supplier: BACHEM AMERICAS INC

4030382.0500 4030382.1000
H-3046.0500BAEA 542.13 CAD
H-3046.0500BA H-3046.1000BA
Bovine;Human;Pig Thr-Ala-Pro-Arg-Atrial Natriuretic Factor (1-28) Trifluoroacetate
Proteins and Peptides

Urodilatin, a 32 amino acid peptide, has originally been isolated from human urine. It belongs to the family of natriuretic-vasorelaxant peptides earlier found in heart atria. Cedidi et al. showed that urodilatin infusion may represent a concept for the treatment of therapy-resistant acute renal failure after liver transplantation. The use of H-3046 is protected by patents. It is sold by Bachem for research purposes only, by expressed permission of Pharis Biotec GmbH, Hannover, Germany (www.pharis-biotec.com).

Order Now

SPECIFICATIONS

Protein/peptide name Thr-Ala-Pro-Arg-Atrial Natriuretic Factor (1-28) Trifluoroacetate
Environmentally Preferable
Protein synonyms Urodilatin|Ularitide
Species Pig,Human,Bovine
Source
CAS No. 115966-23-9
MDL No. MFCD18782181
Sequence H-Thr-Ala-Pro-Arg-Ser-Leu-Arg-Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Met-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-Tyr-OH
Molecular weight 3505.98
Molecular formula C₁₄₅H₂₃₄N₅₂O₄₄S₃
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR