H-Lys-Pro-OH
Supplier: BACHEM AMERICAS INC
4016384.0250
G-4190.0250BAEA 1613.76 CAD
G-4190.0250BA
H-Lys-Pro-OH
Proteins and Peptides
This dipeptide has the potential as a very promising healing agent in ulcer therapy. Daily oral applications to rats accelerated ulcer healing as well by stimulation of angiogenesis as by good mucous regeneration. In mice, a single administration stimulated granulation tissue production.