H-Val-Lys-OH Acetate

Supplier: BACHEM AMERICAS INC

4017637.0250
G-4175.0250BAEA 1161.77 CAD
G-4175.0250BA
H-Val-Lys-OH Acetate
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name H-Val-Lys-OH Acetate
Environmentally Preferable
Species
Source
CAS No. 22677-62-9
MDL No. MFCD00238277
Sequence H-Val-Lys-OH
Molecular weight 245.32
Molecular formula C₁₁H₂₃N₃O₃
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR