3-(3-Pyridyl)-D-alanine dihydrochloride

Supplier: BACHEM AMERICAS INC

4042478.0001
F-4205.0001BAEA 593.81 CAD
F-4205.0001BA
3-(3-Pyridyl)-D-alanine dihydrochloride
3-(3-Pyridyl)-D-alanine dihydrochloride

Sequence: H-β-(3-Pyridyl)-D-Ala-OH · 2 HCl


3-Pal derivatives.


D-3-Pal is a constituent of numerous GnRH antagonists as degarelix, ganirelix, and ozarelix.

Formula: C₈H₁₀N₂O₂·2ClH
MW: 239.099 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00191179
CAS Number: 93960-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR