Ethyl-4-amino-1-methyl-1H-imidazole-2-carboxylate hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: Ethyl 4-amino-1-methyl-1H-imidazole-2-carboxylate hydrochloride, 4-Amino-1-methyl-1H-imidazole-2-carboxylic acid-ethyl ester·HCl

4027071.0001
F-3480.0001BAEA 828.77 CAD
F-3480.0001BA
Ethyl-4-amino-1-methyl-1H-imidazole-2-carboxylate hydrochloride
Ethyl-4-amino-1-methyl-1H-imidazole-2-carboxylate hydrochloride

Sequence: 4-Amino-1-methyl-1H-imidazole-2-carboxylic acid-ethyl ester · HCl


This imidazole derivative can be used in combination with the pyrrole derivative F-3485 for the synthesis of polyamides. Depending on their sequences, these polyamides recognize and bind to specific minor groove DNA sequences.

Formula: C₇H₁₂ClN₃O₂
MW: 205.642 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01318760
CAS Number: 180258-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR