Methyl-2-amino-2-methyl-3-phenylpropanoate hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: Methyl 2-amino-2-methyl-3-phenylpropanoate hydrochloride, H-α-Me-DL-Phe-OMe·HCl

4017873.0005
F-2805.0005BAEA 516.34 CAD
F-2805.0005BA
Methyl-2-amino-2-methyl-3-phenylpropanoate hydrochloride
Methyl-2-amino-2-methyl-3-phenylpropanoate hydrochloride

Sequence: H-α-Me-DL-Phe-OMe · HCl


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.

Formula: C₁₁H₁₆ClNO₂
MW: 229.704 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237767
CAS Number: 64665-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR