H-6-Diazo-5-oxo-D-Nle-OH

Supplier: BACHEM AMERICAS INC

4008357.0025
F-2185.0025BAEA 439.32 CAD
F-2185.0025BA
H-6-Diazo-5-oxo-D-Nle-OH
H-6-Diazo-5-oxo-D-Nle-OH

Sequence: H-6-Diazo-5-oxo-D-Nle-OH
Synonym(s): D-DON#H-D-Glu(diazomethylketone)-OH#6-Diazo-5-oxo-D-2-aminohexanoic acid


Corresponds to D-Glu γ-diazomethylketone.

Formula: C₆H₉N₃O₃
Storage Temperature: Freezer
MDL Number: MFCD00063114
CAS Number: 71629-86-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR