L-Azetidine-2-carboxylic acid

Supplier: BACHEM AMERICAS INC

4019045.0001 4019045.0005
F-1281.0001BAEA 185.3 CAD
F-1281.0001BA F-1281.0005BA
L-Azetidine-2-carboxylic acid
L-Azetidine-2-carboxylic acid

Sequence: L-Azetidine-2-carboxylic acid
Synonym(s): H-Aze-OH


A toxic amino acid found in polygonatum odoratum which acts as a growth inhibitor in plants and algae. It has been incorporated as a proline analog in bioactive peptides for generating conformational changes. Zagari et al. studied its influence on the stability of the collagen helix.

Formula: C₄H₇NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00005166
CAS Number: 2133-34-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR